Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 25kD 34mm flat width (22mm dia meter) 3,7ml/cm including tubing closures and openers wet 1 * 1 m
Catalog # SPEC132127T
Supplier: Spectrum Laboratories
Frequently Bought Together