Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 25kD 28mm flat width (18mm dia meter) 2,5ml/cm including tubing closures and openers wet 1 * 1 m
Katalog # SPEC132126T
Supplier: Spectrum Laboratories
Frequently Bought Together