CELLSTAR®, fles, TC, polystyreen, 50ml, oppervlak 25cm², membraanfilter- schroefdop, 0,2µm, 10st, STERIEL 1 * 200 items
GREI690175RIVM
CELLSTAR®, fles, TC, polystyreen, 50ml, oppervlak 25cm², membraanfilter- schroefdop, 0,2µm, 10st, STERIEL 1 * 200 items
GREI690175RIVM
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection