Order Entry
Denmark
Orders LinkContactUsLinkComponent
RollerTool™ adapters and irradiated covers
RollerTool™ adapters and irradiated covers
PolySorb™ RollerTool™ cover, irradiated, Polyester
Catalog # MICRRTC1112-IR
Supplier:   MICRONOVA
RollerTool™ adapters and irradiated covers
Catalog # MICRRTC1112-IR
Supplier:  MICRONOVA
RollerTool™ adapters and irradiated covers
RollerTool™ adapters and irradiated covers
PolySorb™ RollerTool™ cover, irradiated, Polyester
Price
4.700,00 kr
Log in for availability
1 * 24 Items
Product Details & Documents
A custom mop with dual, curved plates, designed specifically for 3" roller bottle incubators, to clean upper and lower rollers at the same time. Can also be used for small piping and other cylindrical cleanroom surfaces.
  • Clean and sanitize roller bottle incubators with ease
  • Effectively cleans upper and lower rollers at the same time
  • Use with SSU series handles for extended reach inside the roller rack
  • Covers are gamma irradiated for aseptic environments
  • Holds up to strong disinfectants
  • Covers can be laundered for re-use
  • Autoclavable
The Micronova RollerTool™ is made from durable, electropolished stainless steel with a quick disconnect fitting for use with the range of SSU series stainless steel handles. The irradiated covers are available in a choice of fabrics to meet cleaning requirements.
Covers available in a choice of fabrics for specialized cleaning:
NovaPoly™ - smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - textured polyester for residue removal and dispersing disinfectant
MegaTex® - non-woven polyamide is absorbent with a mildly abrasive surface
Specifications
  • Description:
    PolySorb™ RollerTool™ cover, irradiated
  • Material:
    Polyester
  • Seq:
    02