Order Entry
Denmark
Orders LinkContactUsLinkComponent
BIS-TRIS, 0,2 M Buffer Solutions

BIS-TRIS, 0,2 M Buffer Solutions

BIS-TRIS buffer solution, 0,2 M, 7,0, Glass bottle, 250 mL
Catalog # J63346.AK
BIS-TRIS, 0,2 M Buffer Solutions
Catalog # J63346.AK
CAS Number:  
BIS-TRIS, 0,2 M Buffer Solutions
BIS-TRIS, 0,2 M Buffer Solutions

BIS-TRIS buffer solution, 0,2 M, 7,0, Glass bottle, 250 mL

CAS Number:  
Price
1.160,00 kr
Log in for availability
1 * 250 mL
Product Details & Documents

CAS No.: 6976-37-0
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002853

Specifications

Specifications

  • Concentration:
    0,2 M
  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Description:
    BIS-TRIS buffer solution
  • pH value:
    7,0
  • Cat. No.:
    J63346.AK
  • Odour/scent:
    Odourless
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00002853
  • Appearance:
    Colourless
  • CAS No.:
    6976-37-0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.