BIS-TRIS, 0,2 M Buffer Solutions
BIS-TRIS buffer solution, 0,2 M, 6,5, Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 6976-37-0
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00002853
Documents
Specifications
Specifications
- Concentration:0,2 M
- Pack type:Glass bottle
- Pk:250 mL
- Description:BIS-TRIS buffer solution
- pH value:6,5
- Cat. No.:J60127.AK
- Odour/scent:Odourless
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- MDL No.:MFCD00002853
- Appearance:Colourless
- CAS No.:6976-37-0



