Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Human plasma

Human plasma, pooled, 100 ml
Catalog # S4180-100
Supplier:   Biowest
Human plasma
Catalog # S4180-100
Supplier:  Biowest
CAS Number:  
Human plasma
Human plasma

Human plasma, pooled, 100 ml

CAS Number:  
Price
1.140,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

Human plasma is processed from human blood that has been coagulation-induced. It is collected from volunteer donors.

Mainly sourced from France, Germany, Canada or USA, each batch is rigorously controlled and screened for Hepatitis B (HBS), Hepatitis C (HCV) and HIV Types 1 and 2 (HIV 1/2).

 : Products of human origin should be considered potentially infectious and handled accordingly.

Specifications

Specifications

  • Pk:
    100 ml
  • Description:
    Human plasma, pooled

Specifications