Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Human plasma

Human plasma, pooled, 500 ml
Catalog # S4180-500
Supplier:   Biowest
Human plasma
Catalog # S4180-500
Supplier:  Biowest
CAS Number:  
Human plasma
Human plasma

Human plasma, pooled, 500 ml

CAS Number:  
Price
3.290,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Human plasma is processed from human blood that has been coagulation-induced. It is collected from volunteer donors.

Mainly sourced from France, Germany, Canada or USA, each batch is rigorously controlled and screened for Hepatitis B (HBS), Hepatitis C (HCV) and HIV Types 1 and 2 (HIV 1/2).

 : Products of human origin should be considered potentially infectious and handled accordingly.

Specifications

Specifications

  • Pk:
    500 ml
  • Description:
    Human plasma, pooled

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.