Order Entry
Denmark
Orders LinkContactUsLinkComponent
Human sera
Human sera
Human serum AB male, HIV tested, heat inactivated, Sterile filtered, Frozen
Catalog # S419H-100
Supplier:   Biowest
Human sera
Catalog # S419H-100
Supplier:  Biowest
Human sera
Human sera
Human serum AB male, HIV tested, heat inactivated, Sterile filtered, Frozen
Price
2.930,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents
Human serum 'off-the-clot' is processed from human blood that has been coagulation-induced. It is collected from volunteer donors.
Mainly sourced from France, Germany, Canada or USA, each batch is rigorously controlled and screened for Hepatitis B (HBS), Hepatitis C (HCV) and HIV Types 1 and 2 (HIV 1/2).
Important note: Products of human origin should be considered potentially infectious and handled accordingly.
Ordering information: Special treatments are available on request. Please contact your local Avantor Sales Office for more information.
Documents
Specifications
  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    Human serum AB male, HIV tested, heat inactivated
  • Seq:
    03