Order Entry
Denmark
Orders LinkContactUsLinkComponent
Rabbit plasma

Rabbit plasma

Rabbit plasma with EDTA, sterile filtered, 500 ml
Catalog # S2600-500
Supplier:   Biowest
Rabbit plasma
Catalog # S2600-500
Supplier:  Biowest
CAS Number:  
Rabbit plasma

Rabbit plasma with EDTA, sterile filtered, 500 ml

CAS Number:  
Price
924,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Plasma provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth. It is used as a component of bioassays, immunoassays or enzyme assays.

Specifications

Specifications

  • Pk:
    500 ml
  • Description:
    Rabbit plasma with EDTA, sterile filtered

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.