Filter boards
C 160, 142 mm
Catalog # SARTFT-3-343-142
Supplier: Sartorius
Jump to:
Product Details & Documents
Filter boards are used for filtration of cooking oils, transformer oils and as base paper for impregnation with certain reagents.
These are characterised by high wet strength and are therefore especially suitable for pressure or vacuum filtration.
Specifications
- Grade:C 160
- Seq:02
- Size:142 mm
- Thickness:0,30 mm
- Cat. No.:SARTFT-3-343-142
- Wet burst strength:>50 kPa
- Environmental certification:Renewable Energy Certificate
- Capillary rise:80 mm/10 min
- Filtration speed:-
- Environmentally Preferable:Low Manufacturing Impact
- Weight:160 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



