Order Entry
Denmark
Orders LinkContactUsLinkComponent
Filter boards

Filter boards

C 160, 47 mm
Catalog # SARTFT-3-343-047
   Low Manufacturing Impact
Supplier:   Sartorius
Filter boards
Catalog # SARTFT-3-343-047
Supplier:  Sartorius
   Low Manufacturing Impact
Filter boards
Filter boards

C 160, 47 mm

Price
65,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents

Filter boards are used for filtration of cooking oils, transformer oils and as base paper for impregnation with certain reagents.

These are characterised by high wet strength and are therefore especially suitable for pressure or vacuum filtration.

Specifications
  • Grade:
    C 160
  • Seq:
    01
  • Size:
    47 mm
  • Thickness:
    0,30 mm
  • Cat. No.:
    SARTFT-3-343-047
  • Wet burst strength:
    >50 kPa
  • Environmental certification:
    Renewable Energy Certificate
  • Capillary rise:
    80 mm/10 min
  • Filtration speed:
    -
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Weight:
    160 g/m²