Order Entry
Denmark
ContactUsLinkComponent
Disposable Powder Lance HDPE. Blade length 300 mm. Overall length 400 mm. Nominal sample volume 75 mL. Cross Sectional Sampler - No Cleaning Required. Assembled & Packed in a Cleanroom. Individually Bagged. FDA and EU 10/2011 Conforming Materials. BS 1 * 1 items
  SAMP8063H-300S_U
undefined
Disposable Powder Lance HDPE. Blade length 300 mm. Overall length 400 mm. Nominal sample volume 75 mL. Cross Sectional Sampler - No Cleaning Required. Assembled & Packed in a Cleanroom. Individually Bagged. FDA and EU 10/2011 Conforming Materials. BS 1 * 1 items
  SAMP8063H-300S_U
Frequently Bought Together