Order Entry
Denmark
Orders LinkContactUsLinkComponent
Safety spectacles, Rush+
Safety spectacles, Rush+
Anti-UV (2C-1,2), anti-fog, anti-scratch, Red/black, Clear PC
Catalog # 111-1617
Supplier:   Bollé Safety
Safety spectacles, Rush+
Catalog # 111-1617
Supplier:  Bollé Safety
Safety spectacles, Rush+
Safety spectacles, Rush+
Anti-UV (2C-1,2), anti-fog, anti-scratch, Red/black, Clear PC
Price
102,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The RUSH+ model adopts an ultra-sporty design with its co-injected and ultra-flexible temples and is 100% customisable.
  • Upper protection
  • Co-injected, ultra-flexible temples
  • Non-slip adjustable bridge
Available with Twilight technology: ESP with a double anti-fog coating (on both sides of the lenses). With Platinum technology a permanent coating on both sides of the lens gives them a high resistance to scratching (1,4 cd/m²), withstands most aggressive chemicals, and delays the onset of fogging for 2 minutes. Designed for use in low light conditions, it improves contrast. The light transmission rate is ideal for use outdoors, particularly early morning and late evening. B-Twilight filters 76% of blue light.
Certifications: EN 166 FT
Specifications
  • Type:
    Anti-UV (2C-1,2), anti-fog, anti-scratch
  • Colour:
    Red/black
  • Lenses:
    Clear PC