Order Entry
Denmark
ContactUsLinkComponent
Tygon fluoropolymer transfer tubing .25 in od. 5 feet. recommended for elements that may exhibit memory effects with standard nalgen e tubing 1 * 1 items
  VARI190064400
 :  VARIAN
Tygon fluoropolymer transfer tubing .25 in od. 5 feet. recommended for elements that may exhibit memory effects with standard nalgen e tubing 1 * 1 items
  VARI190064400
 :  VARIAN
Frequently Bought Together