Bootie cabinets
Bootie Cabinet 24, 400×400×2100 mm
Catalog # 132-1084
Supplier: Palbam
Jump to:
Product Details & Documents
The modular bootie cabinets come in a variety of shelf lengths.
- Supplied with leveler legs and foot covers
- This symetric product can be used from either side
- Adhesive labelling numbers supplied
Construction: Stainless steel 304L with electropolished finish
Delivery information: Typically available from stock for direct shipment from the supplier.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Description:Bootie Cabinet 24
- W×D×H:400×400×2100 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



