Order Entry
Denmark
ContactUsLinkComponent
VACUUM SSAT-2L (HH) HEPA #111896 Technical Features: Vacuum Pressure: 2032 mm H2O Air Flow: 23,6 L/s Paper Bag Capacity: 1,9 L Dimensions: W21,6 cm x L50,8 cm Weight: 4,99 Kg Accessories Included: #212303: HEPA Filter (H14) #212078SD: Cloth Filter As 1 * 1 Each
Catalog # TIGV111896
Supplier:  TIGER-VAC
VACUUM SSAT-2L (HH) HEPA #111896 Technical Features: Vacuum Pressure: 2032 mm H2O Air Flow: 23,6 L/s Paper Bag Capacity: 1,9 L Dimensions: W21,6 cm x L50,8 cm Weight: 4,99 Kg Accessories Included: #212303: HEPA Filter (H14) #212078SD: Cloth Filter As 1 * 1 Each
Catalog # TIGV111896
Supplier:  TIGER-VAC
Frequently Bought Together