Order Entry
Denmark
ContactUsLinkComponent
Detergent descaler Release 20L 7508888 Concentrated liquid acid detergent Based on citric and phosphoric acid, descaler Light foaming, can be used in CIP Dilute between 1 and 4% w/w Packaging : Canister of 20 liters 1 * 20 l
Catalog # DIVY7517059
Supplier:  Diversey
Detergent descaler Release 20L 7508888 Concentrated liquid acid detergent Based on citric and phosphoric acid, descaler Light foaming, can be used in CIP Dilute between 1 and 4% w/w Packaging : Canister of 20 liters 1 * 20 l
Catalog # DIVY7517059
Supplier:  Diversey
Frequently Bought Together