Spray nozzles
Nozzle for spray gun
Catalog # ENTE215-28
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
PFA spray nozzles with NPT connections for use in applications needing chemical or DI water to be sprayed across a surface.
Molded PFA provides universal chemical compatibility and use in ultrapure and chemically corrosive applications.
Specifications
- Description:Nozzle for spray gun