Order Entry
Denmark
ContactUsLinkComponent
Nitrile glove 30cm TechniGlove TN1202B Class 10 (M2.5), powderfree Length 12´ (300mm) 1 * 1.000 items
Catalog # TCNG107801
Supplier:  TECHNI GLOVE
Nitrile glove 30cm TechniGlove TN1202B Class 10 (M2.5), powderfree Length 12´ (300mm) 1 * 1.000 items
Catalog # TCNG107801
Supplier:  TECHNI GLOVE