Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spermidine

Spermidine

Glass bottle for solids, 10 g
Catalog # CAYM14918-10
Supplier:   Cayman Chemical
Spermidine
Catalog # CAYM14918-10
Supplier:  Cayman Chemical
CAS Number:  124-20-9
Spermidine

Glass bottle for solids, 10 g

CAS Number:  124-20-9
Price
1.860,00 kr
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
  • CAS Number:124-20-9
  • Formula:C₇H₁₉N₃
  • MDL Number:MFCD00008229
  • Molecular Weight:145.25 g/mol
  • Merck Index:12,08893
  • Boiling Pt:246 °C (1013 hPa)
  • Melting Pt:20…25 °C
  • Density:0,925 g/cm³ (20 °C)
  • EINECS:204-689-0
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.