Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bromophenol blue

Bromophenol blue

Glass bottle for solids, 25 g
Catalog # CAYM14331-25
Supplier:   Cayman Chemical
Bromophenol blue
Catalog # CAYM14331-25
Supplier:  Cayman Chemical
CAS Number:  115-39-9
Bromophenol blue

Glass bottle for solids, 25 g

CAS Number:  115-39-9
Price
541,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:13,01429
  • Melting Pt:273 °C
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications