Order Entry
Denmark
ContactUsLinkComponent
Magnifying lamp (table-round base) highly polished glass 1,75X/3X(3 &8-power) lens with large viewing area of 4 . Cover protects lens from dust and scratches. Supplied with 40 LED(2.5W). CH PLUG 1 * 1 items
Catalog # 630-1961
Supplier:  VWR Collection
undefined
Magnifying lamp (table-round base) highly polished glass 1,75X/3X(3 &8-power) lens with large viewing area of 4 . Cover protects lens from dust and scratches. Supplied with 40 LED(2.5W). CH PLUG 1 * 1 items
Catalog # 630-1961
Supplier:  VWR Collection