Fluoride Cell Test Method: photometric 0,10-1,80 mg/L F- Spectroquant, Cell Reagent (Merck Cat. No. 100809) 1 * 1 items
Catalog # WTWA252094
Supplier: WTW
Fluoride Cell Test Method: photometric 0,10-1,80 mg/L F- Spectroquant, Cell Reagent (Merck Cat. No. 100809) 1 * 1 items
Catalog # WTWA252094
Supplier: WTW
Frequently Bought Together
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
pH/ion meter, SevenDirect SD50
Supplier: Mettler - Toledo
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Acetone, for GC Plus, Burdick & Jackson™
Supplier: Honeywell Chemicals
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
n-Octane, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem