Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Single channel pipettes, mechanical, variable volume, VITLAB®

500 - 5000 µl, ±0,6 %, ≤0,2 %, 5000 µl
  VITL1641010
 :   VITLAB
Single channel pipettes, mechanical, variable volume, VITLAB®
  VITL1641010
 :  VITLAB
Single channel pipettes, mechanical, variable volume, VITLAB®
Single channel pipettes, mechanical, variable volume, VITLAB®

500 - 5000 µl, ±0,6 %, ≤0,2 %, 5000 µl

 
3.620,00 kr
   
1 * 1 Items
 

These piston-operated manual pipettes are ideal for demanding laboratory applications.

  • Central, large pipetting button for easy, uniform piston movement
  • Large, ergonomic eject button for rapid replacement of the tips
  • Simple volume adjustment for right- and left-handers
  • Precise, 4-position volume indicator, with integrated zoom function for optimal readability
  • Colour-coded for easy selection of the right pipette tip
  • Ergonomic finger loop for convenient handling
  • Integrated calibration function for easy calibration without the need for tools
  • Corrosion-resistant pistons and ejectors for long product life
  • Completely autoclavable at 121 °C

DE-M marked

 : Supplied with silicone oil resp. grease (Size 5 and 10 ml), sample bag with pipette tips, quality certificate, and instruction manual

 
  • Volume:
    500 - 5000 µl
  • Seq:
    06
  • Accuracy:
    ±0,6 %
  • Imprecision:
    ≤0,2 %
  • Recommended tips:
    5000 µl
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.