Venipuncture sets, Surflo®
Winged needles with protector, G 19, White, 19 mm
Catalog # TERUSV-S19NL30
Supplier: TERUMO
Jump to:
Product Details & Documents
Surshield prevents needle stick injuries with painless placement.
Cannula sharpness combined with a special silicone coating allows an easy and smooth placement.
Specifications
- Gauge:G 19
- Colour:White
- Description:Winged needles with protector
- Length:19 mm