Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cleaning system for controlled environment
Cleaning system for controlled environment
Dip-N-Go Halterung, grey, in stainless steel, 653×622×103 mm
Catalog # VILE143776
Supplier:   FHP VILEDA SCS
Cleaning system for controlled environment
Catalog # VILE143776
Supplier:  FHP VILEDA SCS
Cleaning system for controlled environment
Cleaning system for controlled environment
Dip-N-Go Halterung, grey, in stainless steel, 653×622×103 mm
Price
1.610,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These cleanroom cleaning trolleys are specially designed for use in sensitive areas and controlled environments. The cleaning trolleys are extremely functional, versatile and are characterised by superior ergonomic properties.
  • Small and compact
  • Heat and chemical resistant
  • With electrically conductive castors, the tread of the wheels is made from an hydrolysis resistant material
  • Fully autoclavable and suitable for gamma sterilisation
ISO 5 / GMP A + B / D FED209 class 100
Certifications: ISO 5 / GMP A + B / D FED209 class 100
Specifications
  • Description:
    Dip-N-Go Halterung, grey, in stainless steel
  • Dimensions:
    653×622×103 mm
  • Seq:
    02