Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cleaning system for controlled environment
Cleaning system for controlled environment
CE bucket system trolley, without accessories, suitable for autoclaving and ISO Class 5
Catalog # 141-0937
Supplier:   FHP VILEDA SCS
Cleaning system for controlled environment
Catalog # 141-0937
Supplier:  FHP VILEDA SCS
Cleaning system for controlled environment
Cleaning system for controlled environment
CE bucket system trolley, without accessories, suitable for autoclaving and ISO Class 5
Price
26.070,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These cleanroom cleaning trolleys are specially designed for use in sensitive areas and controlled environments. The cleaning trolleys are extremely functional, versatile and are characterised by superior ergonomic properties.
  • Small and compact
  • Heat and chemical resistant
  • With electrically conductive castors, the tread of the wheels is made from an hydrolysis resistant material
  • Fully autoclavable and suitable for gamma sterilisation
ISO 5 / GMP A + B / D FED209 class 100
Certifications: ISO 5 / GMP A + B / D FED209 class 100
Specifications
  • Description:
    CE bucket system trolley, without accessories, suitable for autoclaving and ISO Class 5
  • Seq:
    12