Order Entry
Denmark
Orders LinkContactUsLinkComponent
Drape sheets, Foliodrape® Protect
Drape sheets, Foliodrape® Protect
Foliodrape® Protect drape sheets, sterile, in 4-layer packaging, 450×750 mm
Catalog # HART277500_U
Supplier:   HARTMANN PAUL
Drape sheets, Foliodrape® Protect
Catalog # HART277500_U
Supplier:  HARTMANN PAUL
Drape sheets, Foliodrape® Protect
Drape sheets, Foliodrape® Protect
Foliodrape® Protect drape sheets, sterile, in 4-layer packaging, 450×750 mm
Price
1.670,00 kr
Log in for availability
1 * 260 Items
Product Details & Documents
These drape sheets are made from a soft polyethylene foil laminated to an absorbent non woven top layer. For sterile draping of patients and equipment during operations with moderate quantities of fluids, especially in outpatient settings.
  • Impermeable to moisture and bacteria
  • Abrasion resistant and extremely low linting
  • Highly conformable
  • Also available as an adhesive drape with self-adhesive area for rapid and accurate positioning at the incision site
Certifications: DIN 13795 (1-3)
Specifications
  • Description:
    Foliodrape® Protect drape sheets, sterile, in 4-layer packaging
  • L×W:
    450×750 mm