Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tissue grinders, CS1, KIMBLE®
Tissue grinders, CS1, KIMBLE®
15 ml
Catalog # SCERSP749600-0015
Supplier:   DWK Life Sciences
Tissue grinders, CS1, KIMBLE®
Catalog # SCERSP749600-0015
Supplier:  DWK Life Sciences
Tissue grinders, CS1, KIMBLE®
Tissue grinders, CS1, KIMBLE®
15 ml
Price
2.380,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
CS1 tissue grinders are suitable for standard grinding, tissues are ground within a sealed container to minimise the risk of personal contact.
  • Narrow pestles eliminate the potential of sample overflow
  • Convenient and easy-to-use
  • Gamma sterilised in an easy-open Tyvek® pack
The molded-in abrasive surface on the pestle tip leaves no sediment to obstruct sample examination. Tubes are made of PP, pestles are 30% glass-filled PP and caps are HDPE.
Delivery information: Each tissue grinder is supplied with a fully assembled pestle, a conical sample tube with solid top cap, an adhesive identification label and an instruction card.
Specifications
  • Capacity:
    15 ml
  • Seq:
    01