Order Entry
Denmark
Orders LinkContactUsLinkComponent
Stirrer blades

Stirrer blades

Moon Shaped blades, PTFE/PEEK, shaft Ø 8 mm, 65×18×3 mm, wrench 15 mm, 24/29 NS
Catalog # BOHLC442-08
Supplier:   Bohlender
Stirrer blades
Catalog # BOHLC442-08
Supplier:  Bohlender
Stirrer blades
Stirrer blades

Moon Shaped blades, PTFE/PEEK, shaft Ø 8 mm, 65×18×3 mm, wrench 15 mm, 24/29 NS

Price
1.600,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Stirrer blades made from PTFE, with a set of clamp screws made from a PTFE/PEEK compound.

  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Create axial flow

The blades can be fixed tightly on stirrer shafts by means of the clamp screws. A spanner wrench is included for easy assembly.

Specifikationer
  • Type:
    Moon Shaped blades, PTFE/PEEK, shaft Ø 8 mm, 65×18×3 mm, wrench 15 mm, 24/29 NS
  • Seq:
    21