Pipette tips, ClickSure™ Tips
Standard, 1000 µl, -, Bulk, 960/bag
Catalog # 613-2935
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
These tips are validated to optimise the performance of the Microlab 300 system, offering superior accuracy and precision. When the tips are placed onto the probe, the O-ring on the probe compresses into the tip groove. This design only requires a low insertion force to successfully load a tip, resulting in a more ergonomic experience for the user and enabling long-term use with minimal energy exerted.
- Tip packaging and tips are recyclable for eco-conscious labs
- Less force is required to load and eject a tip
Specifications
- Packed:Bulk, 960/bag
- Sterile:-
- Volume:1000 µl
- Seq:05
- Tip style:Standard