Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pipette tips, ClickSure™ Tips
Pipette tips, ClickSure™ Tips
Standard, 50 µl, -, Bulk, 960/bag
Catalog # 613-2933
Supplier:   HAMILTON BONADUZ
Pipette tips, ClickSure™ Tips
Catalog # 613-2933
Supplier:  HAMILTON BONADUZ
Pipette tips, ClickSure™ Tips
Pipette tips, ClickSure™ Tips
Standard, 50 µl, -, Bulk, 960/bag
Price
664,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These tips are validated to optimise the performance of the Microlab 300 system, offering superior accuracy and precision. When the tips are placed onto the probe, the O-ring on the probe compresses into the tip groove. This design only requires a low insertion force to successfully load a tip, resulting in a more ergonomic experience for the user and enabling long-term use with minimal energy exerted.
  • Tip packaging and tips are recyclable for eco-conscious labs
  • Less force is required to load and eject a tip
Specifications
  • Packed:
    Bulk, 960/bag
  • Sterile:
    -
  • Volume:
    50 µl
  • Seq:
    01
  • Tip style:
    Standard