Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Docusate sodium USP

Glass, amber, 125 g
Catalog # SPCMDO105-125GM
Irritant
Supplier:   Spectrum Chemical
Docusate sodium USP
Catalog # SPCMDO105-125GM
Supplier:  Spectrum Chemical
CAS Number:  577-11-7
Docusate sodium USP

Glass, amber, 125 g

CAS Number:  577-11-7
Price
1.540,00 kr
Log in for availability
1 * 125 g
Product Details & Documents

Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Synonyms:  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  • CAS Number:577-11-7
  • Formula:C₂₀H₃₇NaO₇S
  • MDL Number:MFCD00012455
  • Molecular Weight:444.57 g/mol
  • Merck Index:12,03460
  • Boiling Pt:∼180 °C (1013 hPa)
  • Melting Pt:∼155 °C
  • Density:∼1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-406-4

 

Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    125 g

Specifications