Diode lasers, class II
Diode laser, blue, Class II, 405 nm
DIDAPOD013136
: DIDALAB
- Power slightly less than 1 mW
- Homogenous and lightly divergent beam (3,2×7,1 mm at 10 metres)
- Electronic device for constant power throughout its use
- Polarized beam - ON / OFF switch
: Laser provided on Ø 10 mm rod with mains power supply.
- Description:Diode laser, blue, Class II
- Wavelength:405 nm



