Diode lasers, class II
Diode laser, red, Class II, 650 nm
Catalog # DIDAPOD013133
Supplier: DIDALAB
Jump to:
Product Details & Documents
- Power slightly less than 1 mW
- Homogenous and lightly divergent beam (3,2×7,1 mm at 10 metres)
- Electronic device for constant power throughout its use
- Polarized beam - ON / OFF switch
Delivery information: Laser provided on Ø 10 mm rod with mains power supply.
Specifications
- Description:Diode laser, red, Class II
- Wavelength:650 nm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



