L-Glutamic acid diethyl ester hydrochloride, 98% 1 * 5 g
H59551.06
Frequently Bought Together
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
: Honeywell Chemicals
Water ACS, Deionized
: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Sinapic acid, Supelco®
: Merck
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
ETHYL ACETATE EXTRA PURE, DAC, N. F. 1 * 100 L
: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
: Honeywell Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Isobutanol 99+% ACS
: Thermo Fisher Scientific
n-Octane ≥98%
: Thermo Fisher Scientific
Tetrabutylammonium dihydrogen phosphate 99% for HPLC
: Thermo Fisher Scientific