Antimony Sb - 100 mg/l Standard for ICP/MS in HNO3/Hf Tr 1 * 100 mL
ACSDM350.2N05FP.L1
: CPACHEM
Frequently Bought Together
pH buffer solutions
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
pH buffers, ready to use
: WTW
Buffer solutions in sachets, perpHect® Pouches, Orion™
: Thermo Orion