Lithium Li - 100 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM330.2NP.L1
Supplier: CPACHEM
Lithium Li - 100 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM330.2NP.L1
Supplier: CPACHEM
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals