Bismuth Bi - 100 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM308.2NP.L1
Supplier: CPACHEM
Bismuth Bi - 100 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM308.2NP.L1
Supplier: CPACHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® ARISTAR®, Cobalt Standard Solution, for ICP, 1000 mg/L Co in dil. Nitric Acid (from Co)
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Triethylene Glycol, Reference Standard
Supplier: USP
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
pH buffer, single-use, SINGLET™
Supplier: Hach
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
4'-Ethoxyacetanilide, Supelco®
Supplier: Merck
Turbidity standards, Stablcal®
Supplier: Hach
ICP METAL STANDARD 31 COMPONENT 1 * 100 mL
Supplier: CPACHEM