Order Entry
Denmark
ContactUsLinkComponent
Calibration gas 25 h2s,100co,2.5%ch4,18%o2,n2 34l 4ga18x25w25h100-34 1 * 1 items
Catalog # BWTE4GA18X25W25H10
Supplier:  BW TECHNOLOGIES
Calibration gas 25 h2s,100co,2.5%ch4,18%o2,n2 34l 4ga18x25w25h100-34 1 * 1 items
Catalog # BWTE4GA18X25W25H10
Supplier:  BW TECHNOLOGIES
Frequently Bought Together