Order Entry
Denmark
ContactUsLinkComponent
Stackable container 300 L, made of High-density polyethylene, not for foods, temperature range: -40° C +80° C and during the washing up to 120 ° C, colour industrial grey, outside dimensions 1000x700x650 cm 1 * 1 items
Catalog # LAMA1432/I
Supplier:  LAMAPLAST
Stackable container 300 L, made of High-density polyethylene, not for foods, temperature range: -40° C +80° C and during the washing up to 120 ° C, colour industrial grey, outside dimensions 1000x700x650 cm 1 * 1 items
Catalog # LAMA1432/I
Supplier:  LAMAPLAST
Frequently Bought Together