Cleanroom boot covers, Kimtech™ A5
Standard vinyl sole, White, S/M, +
Supplier: Ansell
Jump to:
Product Details & Documents
Sterile Kimtech™ A5 cleanroom boot covers are made of SMS (Spunbond Melt-blown Spunbond) material that gives them a cloth-like feeling. Available with standard vinyl sole or with wrap-around vinyl foot.
- Wrap-around vinyl foot is seamed above the floor line to reduce risk of liquid penetration and trip hazards
- Tunnelled elastic and bound seams
- Two extra-long ties
Suitable for use in Class 100, ISO 5, M3.5 environments.
*One size fits all
Certifications: PPE Cat I (EU) 2016/425
Packaging: Vacuum packed in pairs for added sterility assurance.
Specifications
- Shoe size:S/M
- Sterile:+
- Colour:White
- Description:Standard vinyl sole
- Seq:02
- Cat. No.:113-7735
- Cleanroom class:100/ISO 5
- Garment application:Cleanroom
- Environmentally Preferable:Waste Reducing
Complementary Products
Product Information
- Catalog #DescriptionSterileClothing SizeColourLengthAvailabilityPrice
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



