Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cleanroom boot covers, Kimtech™ A5
Standard vinyl sole, White, U*, +
Catalog # 113-7736
   Waste Reducing
Supplier:   Ansell
Cleanroom boot covers, Kimtech™ A5
Catalog # 113-7736
Supplier:  Ansell
   Waste Reducing
Cleanroom boot covers, Kimtech™ A5
Cleanroom boot covers, Kimtech™ A5
Standard vinyl sole, White, U*, +
Price
7.420,00 kr
Log in for availability
1 * 100 Par
Product Details & Documents
Sterile Kimtech™ A5 cleanroom boot covers are made of SMS (Spunbond Melt-blown Spunbond) material that gives them a cloth-like feeling. Available with standard vinyl sole or with wrap-around vinyl foot.
  • Wrap-around vinyl foot is seamed above the floor line to reduce risk of liquid penetration and trip hazards
  • Tunnelled elastic and bound seams
  • Two extra-long ties
Suitable for use in Class 100, ISO 5, M3.5 environments.
*One size fits all
Certifications: PPE Cat I (EU) 2016/425
Emballering: Vacuum packed in pairs for added sterility assurance.
Specifications
  • Shoe size:
    U*
  • Sterile:
    +
  • Colour:
    White
  • Description:
    Standard vinyl sole
  • Seq:
    01
  • Cat. No.:
    113-7736
  • Cleanroom class:
    100/ISO 5
  • Garment application:
    Cleanroom
  • Environmentally Preferable:
    Waste Reducing
Complementary Products
Product Information
  • Catalog #
    Description
    Sterile
    Clothing Size
    Colour
    Length
    Availability
    Price
  • 113-1199
    Without hood
    +
    M
    White
    Log in for availability
    1 * 25 Items - 4.150,00 kr
  • 113-1210
    Without hood
    +
    L
    White
    Log in for availability
    1 * 25 Items - 4.150,00 kr
  • 113-1198
    Without hood
    +
    S
    White
    Log in for availability
    1 * 25 Items - 4.150,00 kr
  • 113-1211
    Without hood
    +
    XL
    White
    Log in for availability
    1 * 25 Items - 4.150,00 kr
  • 113-1212
    Without hood
    +
    XXL
    White
    Log in for availability
    1 * 25 Items - 4.150,00 kr
  • 113-1213
    Without hood
    +
    3XL
    White
    Log in for availability
    1 * 25 Items - 4.150,00 kr
  • 113-1219
    Without hood
    +
    4XL
    White
    Log in for availability
    1 * 25 Items - 4.150,00 kr
  • 113-7732
    With integrated mask
    +
    01
    Log in for availability
    1 * 75 Items - 5.380,00 kr
  • 114-9642
    Standard
    +
    03
    Log in for availability
    1 * 100 Items - 3.360,00 kr
  • 113-7733
    457 mm
    +
    White
    Log in for availability
    1 * 100 Par - 3.710,00 kr