0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Reaction Buffer Phire Green 4x1,5ml 1 * 1 KIT
F-527L
:
FINNZYMES REAGENTS
Reaction Buffer Phire Green 4x1,5ml 1 * 1 KIT
F-527L
:
FINNZYMES REAGENTS
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Carboys, narrow neck, PC, square, Biotainer™, Nalgene®
: Thermo Fisher Scientific
Spare Closures for VWR® Media Bottles
: VWR Collection
Cryoboxes, 133×133 mm
: RATIOLAB
EINWEGMESSPLATTE D-PP25/SS/S07 1 * 1 items
: Anton Paar
Pierce™ Immunoassay Microplates, Steptavidin Coated, 96-Well
: Thermo Fisher Scientific
Sealing mats for assay and storage microplates, Axygen®
: Corning