Reversed phase bulk packing material
PRP-C18 bulk resin, 12 - 20 µm, 100 Å, 1 g
553-0033
Polymer bulk packing material for reversed phase applications.
Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.
- Particle Size:12 - 20 µm
- Pk:1 g
- Pore size:100 Å
- Description:PRP-C18 bulk resin



