Order Entry
Denmark
Orders LinkContactUsLinkComponent
Reversed phase bulk packing material

Reversed phase bulk packing material

PRP-C18 bulk resin, 5 µm, 100 Å, 1 g
Catalog # 553-0031
Supplier:   HAMILTON BONADUZ
Reversed phase bulk packing material
Catalog # 553-0031
Supplier:  HAMILTON BONADUZ
CAS Number:  
Reversed phase bulk packing material
Reversed phase bulk packing material

PRP-C18 bulk resin, 5 µm, 100 Å, 1 g

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents

Polymer bulk packing material for reversed phase applications.

Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.

Documents

Specifications

Specifications

  • Particle Size:
    5 µm
  • Pk:
    1 g
  • Pore size:
    100 Å
  • Description:
    PRP-C18 bulk resin

Specifications