HisPur™ cobalt superflow agarose, Pierce™
HisPur™ Cobalt Superflow Agarose, Liquid, 250 mL
Katalog # PIER25230
Leverandør: Thermo Fisher Scientific
HisPur™ cobalt superflow agarose, Pierce™
Katalog # PIER25230
Leverandør: Thermo Fisher Scientific
CAS nummer:
Gå til:
Product Details & Documents
Pierce™ HisPur™ Cobalt Superflow Agarose is a tetradentate-chelating Superflow 6 support charged with divalent cobalt, designed for high-purity, large-scale purification of polyhistidine-tagged proteins.
- Binds 20 mg of 6xHis-GFP per ml of resin at a linear flow rate of 150 cm/hour; can bind >30 mg per ml of resin depending on flow rate
- Highly crosslinked beads tolerate linear flow rates up to 1200 cm/hour with minimal leaching of cobalt
- Specificity of cobalt binding to histidine-tag generally results in elution fractions with less than 10% contamination of non specific proteins
- Maintains function after exposure to a wide variety of chemicals and pH values
- Resin is stable through multiple cycles of cleaning and reuse
HisPur™ Cobalt Superflow Agarose is a durable resin that does not compress at flow rates used for medium- or large-scale FPLC purifications.
Specifikationer
Specifikationer
- Pk:250 mL
- Description:HisPur™ Cobalt Superflow Agarose
- Type:Liquid
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



