Order Entry
Denmark
ContactUsLinkComponent
Elite-ffap, length: 15m, inner diameter: 0.45mm, film thickness: 0.85µm, temperature limits: 40 c to 250 c. perkinelmer elite-ffap n itroterephthalic acid modified polyetylene glycol (bonded) capillary column, length: 15m, inner diameter: 0.45mm, fil 1 * 1 items
Catalog # PERCN9316355
Supplier:  PerkinElmer
Elite-ffap, length: 15m, inner diameter: 0.45mm, film thickness: 0.85µm, temperature limits: 40 c to 250 c. perkinelmer elite-ffap n itroterephthalic acid modified polyetylene glycol (bonded) capillary column, length: 15m, inner diameter: 0.45mm, fil 1 * 1 items
Catalog # PERCN9316355
Supplier:  PerkinElmer
Frequently Bought Together