Order Entry
Denmark
ContactUsLinkComponent
Each v-21+k wear metals standard 10 µg/g, hydrocarbon oil, 200 g includes the following elements: ag, al, ba, b, cd, ca, cr, cu, fe, k, mg, mn, mo, na, ni, p, pb, si, sn, ti, v, zn concentration: 10 µg/g matrix: hydrocarbon and oilvolume: 200 g. i 1 * 1 items
Catalog # PERCN9308356
Supplier:  PerkinElmer
Each v-21+k wear metals standard 10 µg/g, hydrocarbon oil, 200 g includes the following elements: ag, al, ba, b, cd, ca, cr, cu, fe, k, mg, mn, mo, na, ni, p, pb, si, sn, ti, v, zn concentration: 10 µg/g matrix: hydrocarbon and oilvolume: 200 g. i 1 * 1 items
Catalog # PERCN9308356
Supplier:  PerkinElmer
Frequently Bought Together